missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ZNF324 Polyclonal Antibody
GREENER_CHOICE

Product Code. 17190654 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17190654 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17190654 Supplier Invitrogen™ Supplier No. PA5112798

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

ZNF324 Polyclonal Antibody for Western Blot, IHC (P), ELISA

ZNF324 belongs to the krueppel C2H2-type zinc-finger protein family.It contains 9 C2H2-type zinc fingers and 1 KRAB domain. ZNF324 may be involved in transcriptional regulation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF324
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.38 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene ZNF324
Gene Accession No. O75467
Gene Alias ZF5128; zinc finger protein 324; zinc finger protein 324A; Zinc finger protein ZF5128; ZNF324; ZNF324A
Gene Symbols ZNF324
Host Species Rabbit
Immunogen Recombinant Human Zinc finger protein 324A protein (1-270AA).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25799
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.