missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
ZNF324 Polyclonal antibody specifically detects ZNF324 in Human samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | ZNF324 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Janelia Fluor 646 |
| Dilution | Western Blot |
| Formulation | 50mM Sodium Borate |
| Gene Alias | ZF5128, zinc finger protein 324, zinc finger protein 324A, zinc finger protein ZF5128, ZNF324A |
| Host Species | Rabbit |
| Immunogen | The immunogen for this product maps to a region between residue 503 and 553 of human Zinc Finger Protein 324 using the numbering given in entry NP_055162.1 (GeneID 25799). |
| Purification Method | Immunogen affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?