missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF323 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 25593-1-AP-150UL
This item is not returnable.
View return policy
Description
ZNF323 is a member of the subfamily of C2H2 Kruppel-like zinc finger transcription factors that have a SCAN box domain (Pi et al., 2002 ).Specifications
| ZNF323 | |
| Polyclonal | |
| Unconjugated | |
| ZSCAN31 | |
| zinc finger protein 323, ZNF20 Lp, ZNF310P, ZNF323 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 64288 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.24 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q96LW9 | |
| ZSCAN31 | |
| ZNF323 Fusion Protein Ag22405 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction