missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ZNF323 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA552387
This item is not returnable.
View return policy
Description
Immunogen sequence: EEAVAVVEDL EQELSEPGNQ APDHEHGHSE VLLEDVEHLK VKQEPTDIQL QPMVTQLRYE SFCLHQFQEQ DGESIPENQE LASKQEILKE MEHLGDSKLQ RDVSLDSK Highest antigen sequence identity to the following orthologs: Mouse - 35%, Rat - 31%.
ZNF323 is a member of the subfamily of C2H2 Kruppel-like zinc finger transcription factors that have a SCAN box domain (Pi et al., 2002 ).
Specifications
| ZNF323 | |
| Polyclonal | |
| Unconjugated | |
| ZSCAN31 | |
| dJ874C20.2; zinc finger and SCAN domain containing 31; zinc finger and SCAN domain-containing protein 31; Zinc finger protein 323; ZNF20-Lp; ZNF310P; ZNF323; ZSCAN31 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 64288 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ChIP Assay, Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q96LW9 | |
| ZSCAN31 | |
| Recombinant protein corresponding to Human ZNF323. Recombinant protein control fragment (Product #RP-89443). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction