missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF322A Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-20990
This item is not returnable.
View return policy
Description
ZNF322A Polyclonal specifically detects ZNF322A in Mouse, Rat samples. It is validated for Western Blot.
Specifications
| ZNF322A | |
| Polyclonal | |
| Unconjugated | |
| PBS (pH 7.0), 1.0% BSA and 20% Glycerol with 0.01% Thimerosal | |
| FLJ23393, HCG12, HLA complex group 12, zinc finger protein 322, zinc finger protein 322A, zinc finger protein 388, zinc finger protein 489, ZNF322A, ZNF388, ZNF489 | |
| Rabbit | |
| 47 kDa | |
| 0.1 mL | |
| Primary | |
| Mouse, Rat. Species reactivity based on sequence identity: Human (100%), Bovine (93%). | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
| Western Blot | |
| 0.6mg/mL | |
| Western Blot 1:500-1:3000 | |
| Q6U7Q0 | |
| ZNF322 | |
| Recombinant protein encompassing a sequence within the center region of human ZNF322A. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| 79692 | |
| Mouse, Rat | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction