missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ ZNF319 293T Cell Transient Overexpression Lysate (Denatured)
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
Specifications
Specifications
| Cell Line | 293T |
| For Use With (Application) | Western Blot |
| Description | ZNF319 293T Cell Transient Overexpression Lysate(Denatured) |
| Gene Accession No. | NM_020807.1 |
| Molecular Weight (g/mol) | Theoretical M.W. 64.13kDa |
| Plasmid | pCMV-ZNF319 full-length |
| Quality Control Testing | Transient overexpression cell lysate was tested with anti-ZNF319 antibody (Mfr. No. H00057567-B01) by Western Blot. |
| Quantity | 100 μL |
| Storage Buffer | 1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue) |
| Host Species | Human |
| Afficher plus |
Sécurité et traitement
Identifiant produit
- 100UL ZNF319 293T Cell Transient Overexpression Ly
- Warning
- Skin sensitiser Category 1
- H317-May cause an allergic skin reaction.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P272-Contaminated work clothing should not be allowed out of the workplace.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- MIXTURE LIST-Contains : Mercaptoethanol
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu