Learn More
Invitrogen™ ZNF313 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5120099
Description
Positive test controls include: U-937, HeLa, HepG2, Mouse thymus. The target is usually found in the following locations: Cytoplasm, Nucleus. Immunogen sequence: MAAQQRDCGG AAQLAGPAAE ADPLGRFTCP VCLEVYEKPV QVPCGHVFCS ACLQECLKPK KPVCGVCRSA LAPGVRAVEL ERQIESTETS CHGCRKNFFL SKIRSHVATC SKYQNYIMEG VKATIKDASL QPRNVPNRYT FPCPYCPEKN FDQEGLVEHC KLFHSTDTKS VVCPICASMP WGDPNYRSAN FREHIQRRHR FSYDTFVDYD VDEEDMMNQV LQRSIIDQ.
ZNF313 is a novel human zinc finger protein gene that contains both ring finger and C(2)H(2) domain. It may play a role in spermatogenesis.
Specifications
| ZNF313 | |
| Polyclonal | |
| Unconjugated | |
| RNF114 | |
| 1110008J21Rik; AI225886; AW549494; E3 ubiquitin-protein ligase RNF114; PSORS12; RING finger protein 114; RING-type E3 ubiquitin transferase RNF114; RNF114; Zfp228; Zfp313; Zinc finger protein 228; zinc finger protein 313; ZNF228; Znf313 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 55905, 81018 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 0.47 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9ET26, Q9Y508 | |
| RNF114 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-228 of human RNF114 (NP_061153.1). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.