missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ZNF31 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA553673
This item is not returnable.
View return policy
Description
Immunogen sequence: RSYRKAKSSH PPGTCPFYEE LDSLMRARAA VRAMGTVREA AGLPRCGQSS AETDAQEAWG EVANEDAVKP STLCPKAPDM GFEMRHEDED QISEQDIFEG LPGALSKCPT EAVCQPLDWG EDSENENEDE GQWG Highest antigen sequence identity to the following orthologs: Mouse - 64%, Rat - 61%.
May be involved in transcriptional regulation.
Specifications
| ZNF31 | |
| Polyclonal | |
| Unconjugated | |
| ZSCAN20 | |
| C130001F22; KOX29; Zfp31; ZFP-31; zinc finger and SCAN domain containing 20; zinc finger and SCAN domain-containing protein 20; zinc finger and SCAN domains 20; zinc finger protein 31; Zinc finger protein 360; zinc finger protein KOX29; zinc finger with KRAB and SCAN domains 20; Zkscan20; ZNF31; ZNF360; ZSCAN20 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 7579 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P17040 | |
| ZSCAN20 | |
| Recombinant protein corresponding to Human ZNF31. Recombinant protein control fragment (Product #RP-91317). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction