missing translation for 'onlineSavingsMsg'
Learn More

ZNF307 Antibody (OTI6G6), Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals NBP2-45444

Product Code. 18170148

  • £484.00 / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Ensure accurate, reproducible results in Western Blot, Immunohistochemistry

ZNF307 Monoclonal specifically detects ZNF307 in Human samples. It is validated for Western Blot, Immunohistochemistry.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

ZNF307
Monoclonal
1 mg/mL
Western Blot 1:2000, Immunohistochemistry 1:2000
EC 2.1.1.43, EC 4.2.1.52, FLJ32136, P1P373C6, p373c6.1, Zinc finger protein 307p373c6.1 (novel C2H2 type zinc finger protein), Zinc finger protein 427zinc finger protein with KRAB and SCAN domains 4, zinc finger with KRAB and SCAN domains 4, ZNF307P373c6.1, ZNF427
Mouse
61.4 kDa
0.1 mL
Primary
Human
Purified
Western Blot, Immunohistochemistry
OTI6G6
Unconjugated
PBS (pH 7.3), 1.0% BSA and 50% Glycerol with 0.02% Sodium Azide
ZKSCAN4
Human recombinant protein fragment corresponding to amino acids 1-316 of human ZKSCAN4 (NP_061983) produced in E.coli.
Immunogen affinity purified
RUO
387032
Store at -20C. Avoid freeze-thaw cycles.
IgG1
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only