missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ZNF302 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA567541
This item is not returnable.
View return policy
Description
ZNF302 Polyclonal Antibody for IHC, IHC (P)
May be involved in transcriptional regulation.
Specifications
| ZNF302 | |
| Polyclonal | |
| Unconjugated | |
| ZNF302 | |
| HSD16; MST154; MSTP154; Zinc finger protein 135-like; Zinc finger protein 140-like; Zinc finger protein 302; zinc finger protein 327; ZNF135L; ZNF140L; ZNF302; ZNF327 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 55900 | |
| -20°C | |
| Liquid |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 0.7 mg/mL | |
| PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| Q9NR11 | |
| ZNF302 | |
| Fusion protein of human ZNF302. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction