missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ZNF3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Marca: Invitrogen™ PA540727
Les retours ne sont pas autorisés pour ce produit.
Consulta la politica di reso
Descrizione
Peptide sequence: METQADLVSQ EPQALLDSAL PSKVPAFSDK DSLGDEMLAA ALLKAKSQEL Sequence homology: Cow: 92%; Dog: 100%; Goat: 92%; Horse: 92%; Human: 100%; Mouse: 100%; Pig: 92%; Rat: 100%.
ZNF3 belongs to the krueppel C2H2-type zinc-finger protein family. It contains 8 C2H2-type zinc fingers and 1 KRAB domain. ZNF3 is involved in cell differentiation and/or proliferation.
Specifica
| ZNF3 | |
| Polyclonal | |
| Unconjugated | |
| ZNF3 | |
| A8-51; AI854635; B230309J24; C2-H2 type zinc finger protein; Fnp-1; HF.12; KOX25; LOW QUALITY PROTEIN: zinc finger protein 3; MGC128512 protein; PP838; RGD1565881; Zfp113; Zfp3; Zfp-3; zinc finger and SCAN domain containing 21; zinc finger protein 3; zinc finger protein 3 homolog; zinc finger protein HF.12; Zinc finger protein HZF3.1; zinc finger protein KOX25; ZNF3; ZSCAN21 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 7551 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| P17036 | |
| ZNF3 | |
| Synthetic peptide directed towards the N-terminal of human ZNF3 (aa 1-50). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto