Learn More
ZNF3 Polyclonal Antibody
GREENER_CHOICE
Brand:  Thermo Scientific PA5113628
Product Code. 17181484
  • £437.00 / 100µL
Specifications
ZNF3
Polyclonal
Unconjugated
ZNF3
A8-51; AI854635; B230309J24; C2-H2 type zinc finger protein; Fnp-1; HF.12; KOX25; LOW QUALITY PROTEIN: zinc finger protein 3; MGC128512 protein; PP838; RGD1565881; Zfp113; Zfp3; Zfp-3; zinc finger and SCAN domain containing 21; zinc finger protein 3; zinc finger protein 3 homolog; zinc finger protein HF.12; Zinc finger protein HZF3.1; zinc finger protein KOX25; ZNF3; ZSCAN21
Rabbit
Affinity Chromatography
RUO
7551
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Western Blot
0.5 mg/mL
PBS with 2% sucrose and 0.09% sodium azide
P17036
ZNF3
Synthetic peptide directed towards the N terminal region of human ZFP3.