missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF295 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-74655
This item is not returnable.
View return policy
Description
ZNF295 Polyclonal specifically detects ZNF295 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Western Blot (Negative).
Specifications
| ZNF295 | |
| Polyclonal | |
| Unconjugated | |
| Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide | |
| MGC15716, Zfp414, zinc finger protein 414 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 49854 | |
| Human | |
| IgG |
| Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Western Blot | |
| 1.0 mg/mL | |
| Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:250-1:1000, Immunoprecipitation 2-5ug/mg Lysate, Immunohistochemistry-Paraffin 1:500-1:2000, Western Blot (Negative) | |
| NP_065778.3 | |
| ZNF295 | |
| The immunogen recognized by this antibody maps to a region between residue 875 and 925 of human zinc finger protein 295 using the numbering given in entry NP_065778.3 (GeneID 49854). | |
| 100 μL | |
| Primary | |
| Human. Based on 100% sequence identity, this antibody is predicted to react with Orangutan, Rhesus Monkey, Gorilla, Chimpanzee, White-tufted-ear marmoset, Crab-eating macaque and Northern white-cheeked gibbon. | |
| Store at 4C. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction