missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ ZNF287 293T Cell Transient Overexpression Lysate (Denatured)
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
Specifications
Specifications
| Cell Line | 293T |
| For Use With (Application) | Western Blot |
| Description | ZNF287 293T Cell Transient Overexpression Lysate(Denatured) |
| Gene Accession No. | NM_020653.1 |
| Molecular Weight (g/mol) | Theoretical M.W. 83.05kDa |
| Plasmid | pCMV-ZNF287 full-length |
| Quality Control Testing | Transient overexpression cell lysate was tested with anti-ZNF287 antibody (Mfr. No. H00057336-B01) by Western Blot. |
| Quantity | 100 μL |
| Storage Buffer | 1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue) |
| Host Species | Human |
| Show More |
Sicurezza e movimentazione
Identificativo prodotto
- 100UL ZNF287 293T Cell Transient Overexpression Ly
- Warning
- Skin sensitiser Category 1
- H317-May cause an allergic skin reaction.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P272-Contaminated work clothing should not be allowed out of the workplace.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- MIXTURE LIST-Contains : Mercaptoethanol
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto