missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF286 Antibody (OTI7D2), Alexa Fluor™ 532, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-74935AF532
This item is not returnable.
View return policy
Description
ZNF286 Monoclonal specifically detects ZNF286 in Human samples. It is validated for Western Blot.
Specifications
| ZNF286 | |
| Monoclonal | |
| Alexa Fluor 532 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| ZNF286A | |
| Recombinant protein fragment corresponding to amino acids 1-252 of human ZNF286 (NP_065703) produced in E.coli. | |
| 0.1 mL | |
| 57335.0 | |
| Store at 4°C in the dark. |
| Western Blot | |
| OTI7D2 | |
| Western Blot | |
| MGC149627, zinc finger protein 286, zinc finger protein 286A | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction