missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF284 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | ZNF284 |
|---|---|
| Concentration | 0.19 mg/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16803175
|
Proteintech
24681-1-AP-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16893165
|
Proteintech
24681-1-AP-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ZNF284 may be involved in transcriptional regulation.Specifications
| ZNF284 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse | |
| Q2VY69 | |
| 342909 | |
| ZNF284 Fusion Protein Ag20336 | |
| Primary | |
| -20°C | |
| ZNF284 |
| 0.19 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| zinc finger protein 284, ZNF284, ZNF284L | |
| ZNF284 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title