missing translation for 'onlineSavingsMsg'
Learn More

ZNF281 Antibody, Alexa Fluor™ 594, Novus Biologicals™

Product Code. 18411225 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18411225 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18411225 Supplier Novus Biologicals Supplier No. NBP176551AF594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

ZNF281 Polyclonal antibody specifically detects ZNF281 in Human, Mouse samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF281
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate Alexa Fluor 594
Formulation 50mM Sodium Borate
Gene Alias FLJ20557, zinc finger protein 416
Host Species Rabbit
Immunogen Antibody was raised against a 19 amino acid synthetic peptide near the carboxy terminus of human ZFP281. The immunogen is located within amino acids 600 - 650 of ZFP281. Amino Acid Squence: YSNKSESQKEDPFNIAEPR
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23528
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.