missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ZNF280A Monoclonal Antibody (OTI3G4)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA526399
This item is not returnable.
View return policy
Description
ZNF280A Monoclonal Antibody for Western Blot, IHC (P)
Zinc-finger proteins contain DNA-binding domains and have a wide variety of functions, most of which encompass some form of transcriptional activation or repression. The majority of zinc-finger proteins contain a Krueppel-type DNA binding domain and a KRAB domain, which is thought to interact with KAP1, thereby recruiting histone modifying proteins. SUHW1 (Suppressor of hairy wing homolog 1), also known as ZNF280A (Zinc finger protein 280A), 3'OY11.1, ZNF636 or ZNF280, is a 542 amino acid protein that contains four C2H2-type zinc fingers. Localized to the nucleus, SUHW1 is thought to function as a transcription factor that may mediate transcriptional regulation events.
Specifications
| ZNF280A | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P59817 | |
| ZNF280A | |
| Human recombinant protein fragment corresponding to amino acids 1-333 of ZNF280A produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI3G4 | |
| Unconjugated | |
| ZNF280A | |
| 3'OY11.1; SUHW1; Suppressor of hairy wing homolog 1; zinc finger protein 280; zinc finger protein 280A; Zinc finger protein 636; ZNF280; ZNF280A; ZNF636 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 129025 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction