missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF276 Antibody (PCRP-ZNF276-1A5), PerCP, Novus Biologicals™
Description
ZNF276 Monoclonal antibody specifically detects ZNF276 in Human samples. It is validated for Flow Cytometry
Specifications
Specifications
| Antigen | ZNF276 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | PCRP-ZNF276-1A5 |
| Conjugate | PerCP |
| Formulation | PBS |
| Gene Alias | DNA-binding protein, KIAA1710Zfp46, zinc finger protein 436, ZNF |
| Host Species | Mouse |
| Immunogen | Recombinant full-length human ZNF276 protein (Uniprot: Q8N554) |
| Purification Method | Protein A or G purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?