missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF276 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-03342
This item is not returnable.
View return policy
Description
ZNF276 Polyclonal specifically detects ZNF276 in Human, Mouse samples. It is validated for Western Blot, Immunoprecipitation (Negative).
Specifications
| ZNF276 | |
| Polyclonal | |
| Unconjugated | |
| Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide | |
| DNA-binding protein, KIAA1710Zfp46, zinc finger protein 436, ZNF | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 92822 | |
| Store at 4C. Do not freeze. |
| Western Blot, Immunoprecipitation | |
| 1.0 mg/mL | |
| Western Blot 1:2000 - 1:10000, Immunoprecipitation (Negative) | |
| NP_001106997.1 | |
| ZNF276 | |
| The immunogen recognized by this antibody maps to a region between residue 564 and 614 of human zinc finger protein 276 using the numbering given in entry NP_001106997.1 (GeneID 92822). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction