missing translation for 'onlineSavingsMsg'
Learn More

ZNF276 Antibody, DyLight 405, Novus Biologicals™

Product Code. 18407507 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18407507 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18407507 Supplier Novus Biologicals Supplier No. NBP103342V

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

ZNF276 Polyclonal antibody specifically detects ZNF276 in Human, Mouse samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF276
Applications Western Blot
Classification Polyclonal
Conjugate DyLight 405
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias DNA-binding protein, KIAA1710Zfp46, zinc finger protein 436, ZNF
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 564 and 614 of human zinc finger protein 276 using the numbering given in entry NP_001106997.1 (GeneID 92822).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 92822
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.