missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
ZNF276 Polyclonal antibody specifically detects ZNF276 in Human, Mouse samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | ZNF276 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Alexa Fluor 700 |
| Dilution | Western Blot |
| Formulation | 50mM Sodium Borate |
| Gene Alias | DNA-binding protein, KIAA1710Zfp46, zinc finger protein 436, ZNF |
| Host Species | Rabbit |
| Immunogen | The immunogen recognized by this antibody maps to a region between residue 564 and 614 of human zinc finger protein 276 using the numbering given in entry NP_001106997.1 (GeneID 92822). |
| Purification Method | Immunogen affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?