missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ZNF264 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA598256
This item is not returnable.
View return policy
Description
ZNF264 Polyclonal Antibody for Western Blot, IHC (P), ELISA
ZNF264 gene encodes a zinc finger protein and belongs to the krueppel C2H2-type zinc-finger protein family. Zinc finger proteins are often localized in the nucleus, bind nucleic acids, and regulate transcription.
Specifications
| ZNF264 | |
| Polyclonal | |
| Unconjugated | |
| ZNF264 | |
| KIAA0412; Zinc finger protein 264; ZNF264 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 9422 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 0.25 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O43296 | |
| ZNF264 | |
| Recombinant Human Zinc finger protein 264 protein (1-220AA). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction