missing translation for 'onlineSavingsMsg'
Learn More

ZNF26 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 19839-1-AP-150UL

 View more versions of this product

Product Code. 17791059

  • £349.00 / 150µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

ZNF26 Polyclonal antibody specifically detects ZNF26 in Human samples. It is validated for WB, ELISA
Specifications

Specifications

ZNF26
Polyclonal
Unconjugated
Rabbit
150 μL
7574
Store at -20°C
Liquid
Western Blot, ELISA
550 μg/mL
PBS with 0.02% sodium azide and 50% glycerol pH 7.3.
Antigen affinity purification
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.