missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF259 Antibody (LG1), DyLight 680, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-22448FR
This item is not returnable.
View return policy
Description
ZNF259 Monoclonal specifically detects ZNF259 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
| ZNF259 | |
| Monoclonal | |
| DyLight 680 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| ZNF259 | |
| murine ZPR1 protein | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| LG1 | |
| Western Blot, Immunocytochemistry/Immunofluorescence | |
| ERP1, EZFIT-related protein 1, KIAA1396MGC150657, MGC150658, Z1971, zinc finger protein 471 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| 8882 | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction