missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ZNF257 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA536012
This item is not returnable.
View return policy
Description
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
ZNF257 may be involved in transcriptional regulation.
Specifications
| ZNF257 | |
| Polyclonal | |
| Unconjugated | |
| ZNF257 | |
| BMZF4; BMZF-4; Bone marrow zinc finger 4; zinc finger protein 257; ZNF257 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 113835 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.67 mg/mL | |
| PBS with 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q9Y2Q1 | |
| ZNF257 | |
| Recombinant protein encompassing a sequence within the N-terminus region of human ZNF257. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction