missing translation for 'onlineSavingsMsg'
Learn More

ZNF256 Antibody (3H7), Novus Biologicals™

Product Code. 18330869 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18330869 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18330869 Supplier Novus Biologicals Supplier No. H00010172M01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

ZNF256 Monoclonal antibody specifically detects ZNF256 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF256
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin), Sandwich ELISA
Classification Monoclonal
Clone 3H7
Conjugate Unconjugated
Dilution Western Blot 1:500
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_005764
Gene Alias zinc finger protein 483, zinc finger protein HIT-10, Zinc finger protein with KRAB and SCAN domains 16, ZKSCAN16KIAA1962
Host Species Mouse
Immunogen ZNF256 (NP_005764.2, 521 a.a. ~ 627 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. CNECGKFFSQSSSLIRHRRSHTGERPYECSECWKSFSNHSSLVKHRRVHTGERPYECSECGKSFSQSSNLTNHQRIHSGERPYECSDCGKFFTFNSNLLKHQNVHKG
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10172
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.