missing translation for 'onlineSavingsMsg'
Learn More

ZNF253 Rabbit anti-Human, Polyclonal, Proteintech

Product Code. p-7211530 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
20 μL
150 μL
Unit Size:
150µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16823705 20 μL 20µL
16813705 150 μL 150µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16823705 Supplier Proteintech Supplier No. 251521AP20UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

ZNF253 may function as a transcription factor. Seems to have a transcriptional repression activity.

Specifications

Antigen ZNF253
Applications Western Blot
Classification Polyclonal
Concentration 0.63 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene ZNF253
Gene Accession No. O75346
Gene Alias BMZF 1, BMZF1, Bone marrow zinc finger 1, zinc finger protein 253, Zinc finger protein 411, ZNF253, ZNF411
Gene Symbols ZNF253
Host Species Rabbit
Immunogen ZNF253 Fusion Protein Ag18570
Purification Method Antigen Affinity Chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56242
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.