missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ZNF25 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA537238
This item is not returnable.
View return policy
Description
This antibody detects endogenous protein at a molecular weight of 90 kDa. Purity is >95% by SDS-PAGE.
Zinc-finger proteins contain DNA-binding domains and have a wide variety of functions, most of which encompass some form of transcriptional activation or repression. The majority of zinc-finger proteins contain a Kruppel-type DNA binding domain and a KRAB domain, which is thought to interact with KAP1, thereby recruiting histone modifying proteins. As a member of the krueppel C2H2-type zinc-finger protein family, ZNF25 (Zinc finger protein 25), also known as Zinc finger protein KOX19, is a 456 amino acid nuclear protein that contains one KRAB domain and twelve C2H2-type zinc fingers. The gene encoding ZNF25 maps to a region of human chromosome 10 that has been a loci of interest for multiple endocrine neoplasia type 2A and 2B. There are two isoforms of ZNF25 that are produced as a result of alternative splicing events.
Specifications
| ZNF25 | |
| Polyclonal | |
| Unconjugated | |
| ZNF25 | |
| KOX19; Zfp9; zinc finger protein 25; zinc finger protein 25 (KOX 19); zinc finger protein KOX19; ZNF25 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 219749 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| P17030 | |
| ZNF25 | |
| A synthetic peptide corresponding to residues in human ZNF25. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction