missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF239 Antibody (PCRP-ZNF239-2A10) [Janelia Fluor™ 525], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-24148JF525
This item is not returnable.
View return policy
Description
ZNF239 Monoclonal antibody specifically detects ZNF239 in Human samples. It is validated for Flow Cytometry
Specifications
| ZNF239 | |
| Monoclonal | |
| Janelia Fluor 525 | |
| KIAA1198, zinc finger protein 490 | |
| Recombinant fragment (around aa83-233) of human ZNF239 protein (exact sequence is proprietary) | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry | |
| PCRP-ZNF239-2A10 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 8187 | |
| Store at 4°C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction