missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
ZNF234 Polyclonal specifically detects ZNF234 in Human samples. It is validated for Western Blot.
Specifications
Specifications
| Antigen | ZNF234 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | PBS, 2% Sucrose |
| Gene Alias | HZF4, ZNF234 zinc finger protein 234, ZNF269 |
| Host Species | Rabbit |
| Immunogen | The immunogen is a synthetic peptide directed towards the middle region of human ZNF234 |
| Purification Method | Immunogen affinity purified |
| Quantity | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?