missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
ZNF234 Polyclonal specifically detects ZNF234 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
Specifications
| Antigen | ZNF234 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | FITC |
| Dilution | Immunohistochemistry, Immunohistochemistry-Paraffin |
| Formulation | PBS |
| Gene Alias | HZF4, ZNF234 zinc finger protein 234, ZNF269 |
| Host Species | Rabbit |
| Immunogen | Produced in rabbits immunized with E. coli-derived Human ZNF234 fragment. |
| Purification Method | Antigen and protein A Affinity-purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?