missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF233 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-80192
This item is not returnable.
View return policy
Description
ZNF233 Polyclonal specifically detects ZNF233 in Human samples. It is validated for Western Blot.
Specifications
| ZNF233 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| FLJ38032, zinc finger protein 233 | |
| Rabbit | |
| 77 kDa | |
| 100 μL | |
| Primary | |
| Rat: 79%. | |
| Human, Rat | |
| IgG |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| NP_861421 | |
| ZNF233 | |
| Synthetic peptide directed towards the middle region of human ZNF233. Peptide sequence ERACKCDVYDKGFSQTSQLQAHQRGHSRDKTYKWEVSDRIFNRNSGLHQR. | |
| Affinity purified | |
| RUO | |
| 353355 | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction