missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ZNF23 Monoclonal Antibody (2D3)
GREENER_CHOICE

Product Code. 15538872
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15538872 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15538872 Supplier Invitrogen™ Supplier No. MA520249

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Peptide Sequence: GLQGLQTDIQ TDNDLTKEMY EGKENVSFEL QRDFSQETDF SEASLLEKQQ EVHSAGNIKK EKSNTIDGTV KDETSPVEEC FFSQSSNSYQ CHTITGEQP.

ZNF23 May be involved in transcriptional regulation. May have a role in embryonic development. Belongs to the krueppel C2H2-type zinc-finger protein family.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF23
Applications ELISA, Western Blot
Classification Monoclonal
Clone 2D3
Concentration 0.2-1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene ZNF23
Gene Accession No. P17027
Gene Alias B230354B21Rik; KOX16; kruppel-like zinc finger factor X31; Zfp23; Zfp612; zinc finger protein 23; zinc finger protein 23 (KOX 16); zinc finger protein 32; zinc finger protein 359; Zinc finger protein 612; Zinc finger protein KOX16; ZNF23; ZNF359; ZNF612
Gene Symbols ZNF23
Host Species Mouse
Immunogen ZNF23 (NP_666016.1, 41 a.a. ~ 139 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7571
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.