missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF227 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-05126-100UL
This item is not returnable.
View return policy
Description
ZNF227 Polyclonal specifically detects ZNF227 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| ZNF227 | |
| Polyclonal | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 7770 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry | |
| Unconjugated | |
| KIAA1084, zinc finger protein 507 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 60-230 of human ZNF227 (NP_872296.1). | |
| 100 μL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction