missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ZNF227 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA599787
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total ZNF227.
ZNF227 is a new candidate transcription factor.
Specifications
| ZNF227 | |
| Polyclonal | |
| Unconjugated | |
| ZNF227 | |
| Zinc finger protein 227; ZNF227 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 7770 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q86WZ6 | |
| ZNF227 | |
| A synthesized peptide derived from human ZNF227(Accession Q86WZ6), corresponding to amino acid residues N199-P249. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction