Learn More
Invitrogen™ ZNF226 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5113931
Description
Immunogen sequence: KSFGRSAHLQ AHQKVHTGDK PYKCDECGKG FKWSLNLDMH QRVHTGEKPY For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 93%; Dog: 93%; Guinea Pig: 86%; Horse: 93%; Human: 100%; Mouse: 86%; Pig: 93%; Rabbit: 92%; Rat: 93%; Zebrafish: 85%.
Specifications
| ZNF226 | |
| Polyclonal | |
| Unconjugated | |
| ZNF226 | |
| Kruppel-associated box protein; zinc finger protein 226; ZNF226 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 7769 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9NYT6 | |
| ZNF226 | |
| Synthetic peptide directed towards the C terminal region of human ZNF226 (aa 650-699). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.