missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF22 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17063-1-AP-150UL
This item is not returnable.
View return policy
Description
ZNF22 encodes a protein that is expressed in the epithelial component of the developing tooth organ during early bud and cap stages as well as in osteoblasts of craniofacial bone and the developing tongue.Specifications
| ZNF22 | |
| Polyclonal | |
| Unconjugated | |
| ZNF22 | |
| HKR T1, KOX15, KROX26, Zfp422, Zinc finger protein 22, Zinc finger protein KOX15, Zinc finger protein Krox 26, ZNF22, ZNF422 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 360389, 67255, 7570 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.26 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P17026, Q9ERU2, Q9ERU3 | |
| Znf22 | |
| ZNF22 Fusion Protein Ag10833 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction