missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ZNF22 Polyclonal Antibody

Description
Peptide sequence: TGEKPYQCSE CGKCFSQSSH LRQHMKVHKE EKPRKTRGKN IRVKTHLPSW Sequence homology: Cow: 91%; Dog: 100%; Horse: 100%; Human: 100%; Pig: 100%; Rabbit: 93%.
ZNF22 encodes a protein that is expressed in the epithelial component of the developing tooth organ during early bud and cap stages as well as in osteoblasts of craniofacial bone and the developing tongue.
Specifications
Specifications
| Antigen | ZNF22 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 2% sucrose and 0.09% sodium azide |
| Gene | ZNF22 |
| Gene Accession No. | P17026 |
| Gene Alias | 2900028O21Rik; AU041373; HKR-T1; Kox15; Krox25; Krox-25; Krox-25-2; Krox26; Krox-26; krox-26 protein; krueppel-type zinc finger protein Krox-25; Zfp422; Zinc finger protein 22; zinc finger protein 22 (KOX 15); zinc finger protein 422; Zinc finger protein KOX15; zinc finger protein Krox-25; Zinc finger protein Krox-26; ZNF22; ZNF422 |
| Gene Symbols | ZNF22 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?