missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ZNF217 Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA5142229
This item is not returnable.
View return policy
Description
This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:16,000.
Binds to the promoters of target genes and functions as repressor. Promotes cell proliferation and antagonizes cell death. Promotes phosphorylation of AKT1 at 'Ser-473'.
Specifications
| ZNF217 | |
| Polyclonal | |
| Unconjugated | |
| ZNF217 | |
| ZABC1; Zinc finger protein 217; ZNF217 | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 7764 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Flow Cytometry, Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| O75362 | |
| ZNF217 | |
| Peptide with sequence C-KNAADDSADKVNKN (aa 604-617). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction