missing translation for 'onlineSavingsMsg'
Learn More

ZNF215 Antibody (2C11), Novus Biologicals™

Product Code. 18382129 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18382129 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18382129 Supplier Novus Biologicals Supplier No. H00007762M01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

ZNF215 Monoclonal antibody specifically detects ZNF215 in Human samples. It is validated for Western Blot, ELISA, KnockDown
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF215
Applications Western Blot, ELISA, KnockDown
Classification Monoclonal
Clone 2C11
Conjugate Unconjugated
Formulation In 1x PBS, pH 7.4
Gene Accession No. AAH14538
Gene Alias BAZ 2, BAZ2, BAZ-2, BWSCR2-associated zinc finger protein 2, zinc finger protein 215, zinc finger protein with KRAB and SCAN domains 11
Host Species Mouse
Immunogen ZNF215 (AAH14538, 1 a.a. ~ 304 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MQPLSKLMAISKPRNLSLREQREVLRADMSWQQETNPVVETHDSEASRQKFRHFQYLKVSGPHEALSQLWELCLQWLRPEIHTKKQIIELLVLEQFLAILPEEVRTWVNLQHPNNSKDMVTLIEDVIEMLEDEDMPCKDSALQMGSIKEKMKAGSRTGKPQEPVTFKDVVVEFSKEEWGQLDSAVKNLYRNVMLENFRNLNSLRKAHLLSKPFESLKLESKKKRWIMEKEIPRKTIFDMKSISGEESSHGVIMTRLTESGHPSSDAWKDGVSLLLPRLEYNDAVSAHCNLHLWVQVILLPQPPE
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7762
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.