missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF211 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 25458-1-AP-20UL
This item is not returnable.
View return policy
Description
May be involved in transcriptional regulation.Specifications
| ZNF211 | |
| Polyclonal | |
| Unconjugated | |
| ZNF211 | |
| C2H2 25, CH2H2 25, zinc finger protein 211, Zinc finger protein C2H2 25, ZNF 25, ZNF211, ZNFC25 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 10520 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.33 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q13398 | |
| ZNF211 | |
| ZNF211 Fusion Protein Ag22135 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction