missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ZNF207 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA564139
This item is not returnable.
View return policy
Description
Immunogen sequence: WCWYCNRDFD DEKILIQHQK AKHFKCHICH KKLYTGPGLA IHCMQVHKET IDAVPNAIPG RTDIELEIYG MEGIPEKDMD ERRRLLEQ Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.
ZNF207 is a new candidate transcription factor.
Specifications
| ZNF207 | |
| Polyclonal | |
| Unconjugated | |
| ZNF207 | |
| 49 kDa zinc finger protein; 8430401D15Rik; BUB3-interacting and GLEBS motif-containing protein ZNF207; BUGZ; hBuGZ; Zep; Zfp207; Zinc finger protein 207; Znf207 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 7756 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.4 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O43670 | |
| ZNF207 | |
| Recombinant protein corresponding to Human ZNF207. Recombinant protein control fragment (Product #RP-103533). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction