Learn More
ZNF207 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA530641
Code : PN
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: A549, HeLa, HepG2, HCT116. Predicted reactivity: Xenopus laevis (81%), Rhesus Monkey (100%), Chimpanzee (83%), Bovine (98%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
ZNF207 is a new candidate transcription factor.Specifications
ZNF207 | |
Polyclonal | |
Unconjugated | |
ZNF207 | |
49 kDa zinc finger protein; 8430401D15Rik; BUB3-interacting and GLEBS motif-containing protein ZNF207; BUGZ; hBuGZ; Zep; Zfp207; Zinc finger protein 207; Znf207 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
7756 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
O43670 | |
ZNF207 | |
Recombinant protein fragment corresponding to a region within amino acids 212 and 407 of Human ZNF207 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |