missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
ZNF205 Polyclonal antibody specifically detects ZNF205 in Human samples. It is validated for Immunofluorescence
Specifications
Specifications
| Antigen | ZNF205 |
| Applications | Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Alexa Fluor 350 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | zinc finger protein 205, Zinc finger protein 210, ZNF210Zfp13 |
| Host Species | Rabbit |
| Immunogen | Produced in rabbits immunized with E. coli-derived Human ZNF205 fragment. |
| Purification Method | Antigen and protein A Affinity-purified |
| Quantity | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?