Learn More
Invitrogen™ ZNF202 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5140097
Description
Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: PWVPDIQEPQ ETQEPEILSF TYTGDRSKDE EECLEQEDLS LEDIHRPVLG EPEIHQTPDW EIVFEDNPGR LNERRFGTNI SQVNS Highest antigen sequence indentity to the following orthologs: Mouse - 61%, Rat - 64%.
ZNF202 (Zinc finger protein 202) is a transcriptional repressor of genes affecting the vascular endothelium as well as lipid metabolism and energy homeostasis. Among its targets are structural components of lipoprotein particles (apolipoproteins AIV, CIII, and E), enzymes involved in lipid processing (lipoprotein lipase, lecithin cholesteryl ester transferase), transporters involved in lipid homeostasis (ABCA1, ABCG1), and several genes involved in processes related to energy metabolism and vascular disease.
Specifications
| ZNF202 | |
| Polyclonal | |
| Unconjugated | |
| ZNF202 | |
| Zinc finger protein 202; zinc finger protein with KRAB and SCAN domains 10; ZKSCAN10; ZNF202; ZSCAN42 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 7753 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O95125 | |
| ZNF202 | |
| Recombinant protein corresponding to Human ZNF202. Recombinant protein control fragment (Product #RP-108967). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.