missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF2 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93369-0.02ml
This item is not returnable.
View return policy
Description
ZNF2 Polyclonal antibody specifically detects ZNF2 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| ZNF2 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| A1-5, Zfp661, zinc finger protein 2, zinc finger protein 2 (A1-5), Zinc finger protein 2.2, ZNF661Zinc finger protein 661 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 136-425 of human ZNF2 (NP_066574.2). TEKQSPSGETRKKSLSRDKGLRRRSALSREILTKERHQECSDCGKTFFDHSSLTRHQRTHTGEKPYDCRECGKAFSHRSSLSRHLMSHTGESPYECSVCSKAFFDRSSLTVHQRIHTGEKPFQCNECGKAFFDRSSLTRHQRIHTGESPYECHQCGKAFSQKSILTRHQLIHTGRKPYECNECGKAFYGVSSLNRHQKAHAGDPRYQCNECGKAFFDRSSLTQHQKIHTGDKPYECSECGKAFSQRCRLTRHQRVHTGEKPFECTVCGKVFSSKSSVIQHQRRYAKQGID | |
| 0.02 mL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 7549 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction