missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ZNF197 Monoclonal Antibody (3C11)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA520946
This item is not returnable.
View return policy
Description
Peptide Sequence: IAAEICPHPP TDLVAFNLQD PQHDSPAPEA SALSQEENPR NQLMALMLLT AQPQELVMFE EVSVCFTSEE WACLGPIQRA LYWDVMLENY GNVTSLGYRK YRRQRNK.
ZNF197 product belongs to the zinc finger protein superfamily, members of which are regulatory proteins characterized by nucleic acid-binding zinc finger domains. The encoded protein contains 20 tandemly arrayed C2H2-type zinc fingers, a Kruppel-associated box (KRAB) domain, and a SCAN box. This transcript turns over rapidly and contains 3' UTR AUUUA motifs, which are often a hallmark of rapid turnover. It is overexpressed in some thyroid papillary carcinomas. ZNF197 is located in a cluster of zinc finger genes at 3p21. Two alternatively spliced transcripts encoding different isoforms have been described.
Specifications
| ZNF197 | |
| Monoclonal | |
| 0.2-1.0 mg/mL | |
| PBS with no preservative; pH 7.4 | |
| O14709 | |
| ZNF197 | |
| ZNF197 (NP_001020026.1, 161 a.a. ~ 267 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 κ |
| ELISA, Western Blot | |
| 3C11 | |
| Unconjugated | |
| ZNF197 | |
| D3S1363E; P18; pVHL-associated KRAB domain-containing protein; VHLaK; VHL-associated KRAB-A domain-containing protein; zinc finger protein 166; zinc finger protein 197; Zinc finger protein 197-like protein; zinc finger protein 20; zinc finger protein with KRAB and SCAN domains 9; ZKSCAN9; ZNF166; ZNF197; ZnF20; ZSCAN41 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 10168 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction