missing translation for 'onlineSavingsMsg'
Learn More

ZNF197 Antibody (3C11), Novus Biologicals™

Product Code. 18359539 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18359539 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18359539 Supplier Novus Biologicals Supplier No. H00010168M15

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

ZNF197 Monoclonal antibody specifically detects ZNF197 in Human samples. It is validated for Western Blot, ELISA, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF197
Applications Western Blot, ELISA, Sandwich ELISA
Classification Monoclonal
Clone 3C11
Conjugate Unconjugated
Dilution Western Blot 1:500
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_001020026
Gene Alias D3S1363E, P18, VHL-associated KRAB-A domain-containing protein, zinc finger protein 166, zinc finger protein 197, zinc finger protein 20, Zinc finger protein with KRAB and SCAN domains 9, ZKSCAN9VHLaK, ZNF166, ZNF20
Host Species Mouse
Immunogen ZNF197 (NP_001020026.1, 161 a.a. ~ 267 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. IAAEICPHPPTDLVAFNLQDPQHDSPAPEASALSQEENPRNQLMALMLLTAQPQELVMFEEVSVCFTSEEWACLGPIQRALYWDVMLENYGNVTSLGYRKYRRQRNK
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10168
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.