missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF195 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 24861-1-AP-150UL
This item is not returnable.
View return policy
Description
ZNF195 contains 1 KRAB domain and 10 C2H2-type zinc fingers and belongs to the krueppel C2H2-type zinc-finger protein family. It may be involved in transcriptional regulation.Specifications
| ZNF195 | |
| Polyclonal | |
| Unconjugated | |
| ZNF195 | |
| zinc finger protein 195, ZNF195, ZNFP104 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 7748 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.35 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| O14628 | |
| ZNF195 | |
| ZNF195 Fusion Protein Ag21735 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction